Tisk…

Anti-non-intact Vtg Mouse Monoclonal Antibody [clone: ND-3G2]

Dodavatel: Cayman Chemical
170107-100 170107-500
CAYM170107-100EA 15200 CZK
CAYM170107-100 CAYM170107-500
Anti-non-intact Vtg Mouse Monoclonal Antibody [clone: ND-3G2]
Protilátky
Type: Primary
Antigen: non-intact Vtg
Clonality: Monoclonal
Clone: ND-3G2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Flounder, large-mouth Bass, Rainbow Trout, striped Bass, Turbot, Wrasse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR