Tisk…

Anti-Cu/Zn SOD Rabbit Polyclonal Antibody

Dodavatel: Cayman Chemical
10011388-100 10011388-25
CAYM10011388-100EA 20930 CZK
CAYM10011388-100 CAYM10011388-25
Anti-Cu/Zn SOD Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: Cu/Zn SOD
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Dog, coral, Hamster, Human, Monkey, Mouse, Sheep, Pig, Rabbit, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR