Tisk…

Anti-Bim Rabbit Polyclonal Antibody

Dodavatel: Cayman Chemical
10011385-100 10011385-25
CAYM10011385-100EA 14560 CZK
CAYM10011385-100 CAYM10011385-25
Anti-Bim Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: Bim
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope: Internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Produktová specifikace

Symbol antigenu BIM
Environmentally Preferable
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Unconjugated
Reaktivita Human,Rat,Mouse
Hostitel Rabbit
Izotyp IgG
Western blot Yes
Imunochemie Yes
Epitop Internal
Imunogen internal central human Bim amino acids
Čištění purified by immunoaffinity chromatography
Křížová adsorbce No
Skladovací pufr PBS containing 0,02% sodium azide
Skladovací teplota –20 °C
Transportní teplota –20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR