Tisk…
Anti-Trail Rabbit Polyclonal Antibody
Dodavatel: US Biological
143684 143684-100
USBI143684EA 9230 CZK
USBI143684 USBI143684-100
Anti-Trail Rabbit Polyclonal Antibody
Protilátky
Recommended dilution: ELISA: 0.5-2?g/ml
Type: Primary
Antigen: Trail
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse,