Tisk…

Anti-Vascular Endothelial Growth Factor A Mouse Polyclonal Antibody

Dodavatel: US Biological
142766 142766-100
USBI142766EA 11990 CZK
USBI142766 USBI142766-100
Anti-Vascular Endothelial Growth Factor A Mouse Polyclonal Antibody
Protilátky
Recommended dilution: ELISA: 1:100-200; Western Blot: 1:100-400; Immunochemistry: 1:100-500

Type: Primary
Antigen: Vascular Endothelial Growth Factor A
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human,
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR