Tisk…

Anti-Atrial Natriuretic Peptide, pro- Mouse Monoclonal Antibody [clone: A136-4-L1-23-G7
]

Dodavatel: US Biological
A4150-03D-10 A4150-03D-100
USBIA4150-03D-10EA 12500 CZK
USBIA4150-03D-10 USBIA4150-03D-100
Anti-Atrial Natriuretic Peptide, pro- Mouse Monoclonal Antibody [clone: A136-4-L1-23-G7
]
Protilátky
Type: Primary
Antigen: Atrial Natriuretic Peptide, pro-
Clonality: Monoclonal
Clone: A136-4-L1-23-G7

Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human,
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR