Tisk…

Anti-Tumor Necrosis Factor alpha Goat Polyclonal Antibody (Biotin)

Dodavatel: US Biological
T9160-28C-25 T9160-28C-50
USBIT9160-28C-25EA 14410 CZK
USBIT9160-28C-25 USBIT9160-28C-50
Anti-Tumor Necrosis Factor alpha Goat Polyclonal Antibody (Biotin)
Protilátky
Recommended dilution: ELISA: 0.15-0. 30 ?g/ml

Type: Primary
Antigen: Tumor Necrosis Factor alpha
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse,
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR