Tisk…
Analytical HPLC columns, Microsorb™ C18
Dodavatel: VARIAN
R0086203C5 R0089200D5 R0086200C5 R0086200D5
VARIR0086203C5EA 24240 CZK
VARIR0086203C5 VARIR0089200D5 VARIR0086200C5 VARIR0086200D5
Analytical HPLC columns, Microsorb™ C18
Kolony chromatografické
Microsorb™ C18 is an octadecylsilane-covered silica that has been bonded using monofunctional bonding agents under optimised conditions. This material is then endcapped with trimethylsilyl groups to minimise nonspecific binding, maximise mass recoveries and improve peak shape.
C18 columns are typically used with water/methanol or water/acetonitrile solvent combinations and are ideal for the separation of small molecules, proteins, or peptides with differing hydrophobicities.