Tisk…
Bottles, narrow neck, amber glass, Boston type with caps
Dodavatel: EP SCIENTIFIC
145-4000 145-2360 345-2360 345-4000 245-4000 245-2360
EPSC145-4000EA 4690 CZK
EPSC145-4000 EPSC145-2360 EPSC345-2360 EPSC345-4000 EPSC245-4000 EPSC245-2360
Bottles, narrow neck, amber glass, Boston type with caps
Láhve Láhve na odběr vzorků vody
Type III soda-lime amber glass, with screw cap.
  • Excellent chemical resistance
  • Good temperature resistance
Designed for collecting water samples for a variety of organic and inorganic target analytes where light protection is required.
Certifikace: Certified to meet EPA performance based specifications for semi-volatile organics, pesticides, PCBs and metals analyses. Available Certified to meet EPA performance-based specifications for semi-volatile organics, pesticides, PCBs and metals analyses, Processed the same way as the certified jugs but without certification documentation, or Unprocessed and ready for your own cleaning procedure.