Tisk…
Capillary columns, DB-FFAP
Dodavatel: Agilent
123-3234 122-3232LTM 123-3232LTM 123-3233LTM 123-3234LTM 125-3217LTM 127-3212LTM 127-32H2LTM
HEWL123-3234EA 27380 CZK
HEWL123-3234 HEWL122-3232LTM HEWL123-3232LTM HEWL123-3233LTM HEWL123-3234LTM HEWL125-3217LTM HEWL127-3212LTM HEWL127-32H2LTM
Capillary columns, DB-FFAP
Kolony chromatografické
These columns are designed for the analysis of volatile fatty acids and phenols.
  • Highly polar
  • Bonded and cross linked
  • Solvent rinsable
Nitroterephthalic acid modified polyethylene glycol.
Max. temperature: 250 °C
Suitable replacement for OV-351.