Tisk…
Láhve odsávací
Dodavatel: GLASSCO
259.245.04 260.245.04
GLAS259.245.04EA 0 CZK
GLAS259.245.04 GLAS260.245.04
Láhve odsávací
Baňky Baňky filtrační
Flasks are made of heat resistant borosilicate glass 3.3, therefore they are suitable for hot and cold filtration.
  • Highly resistant against chemicals and thermal shock
  • High temperature resistance
  • With white inscription field
Due to the heavy wall thickness the VWR® filtration flask is vacuum tight.