Tisk…

Anti-RP1 Rabbit Polyclonal Antibody

Dodavatel: Novus Biologicals
NBP2-22149 NBP2-22149SS
NOVUNBP2-22149EA 15780 CZK
NOVUNBP2-22149 NOVUNBP2-22149SS
Anti-RP1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: RP1 (retinitis pigmentosa 1 (autosomal dominant))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR