Tisk…

Anti-PRKDC Mouse Monoclonal Antibody [clone: 3H6]

Dodavatel: Novus Biologicals
NBP2-22128 NBP2-22128SS
NOVUNBP2-22128EA 15780 CZK
NOVUNBP2-22128 NOVUNBP2-22128SS
Anti-PRKDC Mouse Monoclonal Antibody [clone: 3H6]
Protilátky
Type: Primary
Antigen: PRKDC (protein kinase, DNA-activated, catalytic polypeptide)
Clonality: monoclonal
Clone: 3H6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR