Tisk…
Anti-SETD8 Rabbit Polyclonal Antibody
Dodavatel: Novus Biologicals
NBP1-91269 NBP1-91269SS
NOVUNBP1-91269EA 15780 CZK
NOVUNBP1-91269 NOVUNBP1-91269SS
Anti-SETD8 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: SETD8 (SET domain containing (lysine methyltransferase) 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human