Tisk…

Anti-ILVBL Rabbit Polyclonal Antibody

Dodavatel: Novus Biologicals
NBP1-78433 NBP1-78433SS
NOVUNBP1-78433EA 12700 CZK
NOVUNBP1-78433 NOVUNBP1-78433SS
Anti-ILVBL Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: ILVBL (ilvB (bacterial acetolactate synthase)-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR