Tisk…

Anti-EHMT1 Rabbit Polyclonal Antibody

Dodavatel: Novus Biologicals
NBP1-77400 NBP1-77400SS
NOVUNBP1-77400EA 0 CZK
NOVUNBP1-77400 NOVUNBP1-77400SS
Anti-EHMT1 Rabbit Polyclonal Antibody
Protilátky
The GLP / EHMT1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to GLP / EHMT1. This antibody reacts with human, mouse. The GLP / EHMT1 Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:150

Type: Primary
Antigen: GLP/EHMT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR