Tisk…

Anti-KDM3A Rabbit Polyclonal Antibody

Dodavatel: Novus Biologicals
NBP1-49601 NBP1-49601SS
NOVUNBP1-49601EA 16830 CZK
NOVUNBP1-49601 NOVUNBP1-49601SS
Anti-KDM3A Rabbit Polyclonal Antibody
Protilátky
Rabbit Polyclonal JMJD1A Antibody. Tested Applications: Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human, Mouse, Primate.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:50

Type: Primary
Antigen: JMJD1A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Objednat

Produktová specifikace

Symbol antigenu JMJD1A
Environmentally Preferable
Název antigenu Lysine-specific demethylase 3A
Synonyma antigenu JHDM2A|TSGA|JHMD2A|JMJD1A|JMJD1
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Unconjugated
Reaktivita Human
Hostitel Rabbit
Identifikační číslo genu 55818
Izotyp
Western blot Yes
ImunoFluorescence Yes
Skladovací teplota Cool
Transportní teplota Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR