Tisk…
Anti-RBBP6 Rabbit Polyclonal Antibody
Dodavatel: Novus Biologicals
NBP1-49535 NBP1-49535SS
NOVUNBP1-49535EA 16180 CZK
NOVUNBP1-49535 NOVUNBP1-49535SS
Anti-RBBP6 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: RBBP6 (retinoblastoma binding protein 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human