Tisk…

Anti-LOX propeptide Rabbit Polyclonal Antibody

Dodavatel: Novus Biologicals
NBP1-30327 NBP1-30327SS
NOVUNBP1-30327EA 15780 CZK
NOVUNBP1-30327 NOVUNBP1-30327SS
Anti-LOX propeptide Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: LOX propeptide
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR