Tisk…
Anti-KDM6B Rabbit Polyclonal Antibody
Dodavatel: Novus Biologicals
NBP1-06640 NBP1-06640SS
NOVUNBP1-06640EA 18740 CZK
NOVUNBP1-06640 NOVUNBP1-06640SS
Anti-KDM6B Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: KDM6B (lysine (K)-specific demethylase 6B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse