Tisk…

Anti-HIF1A Rabbit Polyclonal Antibody

Dodavatel: Novus Biologicals
NBP1-02160 NBP1-02160SS
NOVUNBP1-02160EA 0 CZK
NOVUNBP1-02160 NOVUNBP1-02160SS
Anti-HIF1A Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: HIF1A (hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR