Tisk…
Anti-FOXP3 Rabbit Polyclonal Antibody
Dodavatel: Novus Biologicals
NB600-246 NB600-246SS
NOVUNB600-246EA 0 CZK
NOVUNB600-246 NOVUNB600-246SS
Anti-FOXP3 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: FOXP3 (forkhead box P3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse