Tisk…

Anti-NUMB Rabbit Polyclonal Antibody

Dodavatel: Novus Biologicals
NB500-178 NB500-178SS
NOVUNB500-178EA 15780 CZK
NOVUNB500-178 NOVUNB500-178SS
Anti-NUMB Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: NUMB (numb homolog (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Chicken, Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR