Tisk…
Anti-HIST3H3 Rabbit Polyclonal Antibody
Dodavatel: Novus Biologicals
NB21-1023 NB21-1023SS
NOVUNB21-1023EA 0 CZK
NOVUNB21-1023 NOVUNB21-1023SS
Anti-HIST3H3 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: HIST3H3 (histone cluster 3, H3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat