Tisk…

Anti-HIST3H3 Rabbit Polyclonal Antibody

Dodavatel: Novus Biologicals
NB21-1001 NB21-1001SS
NOVUNB21-1001EA 17430 CZK
NOVUNB21-1001 NOVUNB21-1001SS
Anti-HIST3H3 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: HIST3H3 (histone cluster 3, H3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR