Tisk…

Anti-ITGAM Rabbit Polyclonal Antibody

Dodavatel: Novus Biologicals
NB110-89474SS NB110-89474
NOVUNB110-89474SSEA 8750 CZK
NOVUNB110-89474SS NOVUNB110-89474
Anti-ITGAM Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: ITGAM (integrin, alpha M (complement component 3 receptor 3 subunit))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR