Tisk…

Anti-Hepatic Sinusoidal Endothelial Cells Mouse Monoclonal Antibody [clone: SE-1]

Dodavatel: Novus Biologicals
NB110-68095 NB110-68095SS
NOVUNB110-68095EA 18280 CZK
NOVUNB110-68095 NOVUNB110-68095SS
Anti-Hepatic Sinusoidal Endothelial Cells Mouse Monoclonal Antibody [clone: SE-1]
Protilátky
Type: Primary
Antigen: Hepatic Sinusoidal Endothelial Cells
Clonality: monoclonal
Clone: SE-1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR