Tisk…

Anti-SOX2 Rabbit Polyclonal Antibody

Dodavatel: Novus Biologicals
NB110-37235 NB110-37235SS
NOVUNB110-37235EA 19790 CZK
NOVUNB110-37235 NOVUNB110-37235SS
Anti-SOX2 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: SOX2 (SRY (sex determining region Y)-box 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR