Tisk…

Anti-JARID2 Rabbit Polyclonal Antibody

Dodavatel: Novus Biologicals
NB100-2214 NB100-2214SS
NOVUNB100-2214EA 16600 CZK
NOVUNB100-2214 NOVUNB100-2214SS
Anti-JARID2 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: JARID2 (jumonji, AT rich interactive domain 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR