Tisk…

Anti-Sir2.1 Rabbit Polyclonal Antibody

Dodavatel: Novus Biologicals
NB100-1923 NB100-1923SS
NOVUNB100-1923EA 16920 CZK
NOVUNB100-1923 NOVUNB100-1923SS
Anti-Sir2.1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: Sir2.1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: C. elegans
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR