Tisk…

Anti-NCL Rabbit Polyclonal Antibody

Dodavatel: Novus Biologicals
NB100-1920 NB100-1920SS
NOVUNB100-1920EA 16830 CZK
NOVUNB100-1920 NOVUNB100-1920SS
Anti-NCL Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: NCL (nucleolin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR