Tisk…

Anti-ATM Rabbit Polyclonal Antibody

Dodavatel: Novus Biologicals
NB100-104 NB100-104SS
NOVUNB100-104EA 18200 CZK
NOVUNB100-104 NOVUNB100-104SS
Anti-ATM Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: ATM (ataxia telangiectasia mutated)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR