Tisk…

Anti-IgG2a Goat Polyclonal Antibody (CF™ 543)

Dodavatel: Biotium
20326 20326-1
20326.EA 10960 CZK
20326. 20326-1
Anti-IgG2a Goat Polyclonal Antibody (CF™ 543)
Protilátky
Type: Secondary
Antigen: IgG2a (Mouse) (mouse IgG2a)
Clonality: polyclonal
Clone:
Conjugation: CF® 543 Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR