Tisk…

Anti-IgG Donkey Polyclonal Antibody (CF™ 543)

Dodavatel: Biotium
20320 20320-1
20320.EA 10230 CZK
20320. 20320-1
Anti-IgG Donkey Polyclonal Antibody (CF™ 543)
Protilátky
Type: Secondary
Antigen: IgG (Rat) (rat IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 543 Dye
Epitope:
Host: Donkey
Isotype: none
Reactivity: Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR