Tisk…

Anti-IgG Chicken Polyclonal Antibody (CF™ 488A)

Dodavatel: Biotium
20209 20209-1
20209.EA 8530 CZK
20209. 20209-1
Anti-IgG Chicken Polyclonal Antibody (CF™ 488A)
Protilátky
Type: Secondary
Antigen: IgG (Rabbit) (rabbit IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 488A Dye
Epitope:
Host: Chicken
Isotype: none
Reactivity: Rabbit
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR