Tisk…

Anti-IgG Donkey Polyclonal Antibody (Biotin)

Dodavatel: Biotium
20191 20191-1
20191.EA 10230 CZK
20191. 20191-1
Anti-IgG Donkey Polyclonal Antibody (Biotin)
Protilátky
Type: Secondary
Antigen: IgG (Rabbit) (rabbit IgG)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Donkey
Isotype: none
Reactivity: Rabbit
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR