Tisk…

Anti-IgG Rabbit Polyclonal Antibody (CF™ 633)

Dodavatel: Biotium
20136 20136-1
20136.EA 9870 CZK
20136. 20136-1
Anti-IgG Rabbit Polyclonal Antibody (CF™ 633)
Protilátky
Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 633 Dye
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR