Tisk…

Anti-IgG Donkey Polyclonal Antibody (CF™ 633)

Dodavatel: Biotium
20134 20134-1
20134.EA 10230 CZK
20134. 20134-1
Anti-IgG Donkey Polyclonal Antibody (CF™ 633)
Protilátky
Type: Secondary
Antigen: IgG (Sheep) (sheep IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 633 Dye
Epitope:
Host: Donkey
Isotype: none
Reactivity: Sheep
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR