Tisk…

Anti-IgG Goat Polyclonal Antibody (CF™ 405S)

Dodavatel: Biotium
20082 20082-1
20082.EA 8230 CZK
20082. 20082-1
Anti-IgG Goat Polyclonal Antibody (CF™ 405S)
Protilátky
Type: Secondary
Antigen: IgG (Rabbit) (rabbit IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® CF405S Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Rabbit
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR