Tisk…

Anti-IgG Goat Polyclonal Antibody (CF™ 680)

Dodavatel: Biotium
20069 20069-1
20069.EA 9900 CZK
20069. 20069-1
Anti-IgG Goat Polyclonal Antibody (CF™ 680)
Protilátky
Type: Secondary
Antigen: IgG (Rat) (rat IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 680 Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR