Tisk…

Anti-IgG Rabbit Polyclonal Antibody (CF™ 680)

Dodavatel: Biotium
20061 20061-1
20061.EA 9570 CZK
20061. 20061-1
Anti-IgG Rabbit Polyclonal Antibody (CF™ 680)
Protilátky
Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 680 Dye
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR