Tisk…

Anti-IgG Goat Polyclonal Antibody (CF™ 660R)

Dodavatel: Biotium
20055 20055-1
20055.EA 8530 CZK
20055. 20055-1
Anti-IgG Goat Polyclonal Antibody (CF™ 660R)
Protilátky
Type: Secondary
Antigen: IgG (Rabbit) (rabbit IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 660R Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Rabbit
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR