Tisk…

Anti-IgG Goat Polyclonal Antibody (CF™ 555)

Dodavatel: Biotium
20033 20033-1
20033.EA 8530 CZK
20033. 20033-1
Anti-IgG Goat Polyclonal Antibody (CF™ 555)
Protilátky
Type: Secondary
Antigen: IgG (Rabbit) (rabbit IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 555 Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Rabbit
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR