Tisk…
Anti-HSPC150/UBE2T Rabbit Monoclonal Antibody [clone: EPR9446]
Dodavatel: Abcam
AB140611-100 AB140611-1ML
ABCAAB140611-100EA 20130 CZK
ABCAAB140611-100 ABCAAB140611-1ML
Anti-HSPC150/UBE2T Rabbit Monoclonal Antibody [clone: EPR9446]
Protilátky
Rabbit monoclonal [EPR9446] to HSPC150/UBE2T.
Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: HSPC150/UBE2T
Clonality: Monoclonal
Clone: EPR9446
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human