Tisk…

Anti-HLA Class II DRB1 Rabbit Monoclonal Antibody [clone: EPR6148]

Dodavatel: Abcam
AB133578-100 AB133578-20UL AB133578-1ML
ABCAAB133578-100EA 18890 CZK
ABCAAB133578-100 ABCAAB133578-20UL ABCAAB133578-1ML
Anti-HLA Class II DRB1 Rabbit Monoclonal Antibody [clone: EPR6148]
Protilátky
Rabbit monoclonal [EPR6148] to HLA Class II DRB1.

Upozornení: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: HLA Class II DRB1
Clonality: Monoclonal
Clone: EPR6148
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR