Tisk…

Anti-Angiotensin II Type 1 Receptor Rabbit Monoclonal Antibody [clone: EPR3873]

Dodavatel: Abcam
AB124734-100 AB124734-1ML
ABCAAB124734-100EA 19420 CZK
ABCAAB124734-100 ABCAAB124734-1ML
Anti-Angiotensin II Type 1 Receptor Rabbit Monoclonal Antibody [clone: EPR3873]
Protilátky
Rabbit monoclonal [EPR3873] to Angiotensin II Type 1 Receptor.

Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Angiotensin II Type 1 Receptor
Clonality: Monoclonal
Clone: EPR3873
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR