Tisk…

Anti-PAP Rabbit Monoclonal Antibody [clone: EPR4066]

Dodavatel: Abcam
AB108984-100 AB108984-1ML
ABCAAB108984-100EA 20130 CZK
ABCAAB108984-100 ABCAAB108984-1ML
Anti-PAP Rabbit Monoclonal Antibody [clone: EPR4066]
Protilátky
Rabbit monoclonal [EPR4066] to PAP.

Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: PAP
Clonality: Monoclonal
Clone: EPR4066
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR