Tisk…
Anti-Vitamin D Binding protein Rabbit Monoclonal Antibody [clone: EP2923Y]
Dodavatel: Abcam
AB81307-100 AB81307-1ML
ABCAAB81307-100EA 18890 CZK
ABCAAB81307-100 ABCAAB81307-1ML
Anti-Vitamin D Binding protein Rabbit Monoclonal Antibody [clone: EP2923Y]
Protilátky
Rabbit monoclonal [EP2923Y] to Vitamin D Binding protein.
Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Vitamin D Binding protein
Clonality: Monoclonal
Clone: EP2923Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human