Tisk…

Anti-delta 1 Catenin/CAS (phospho S268) Rabbit Monoclonal Antibody [clone: EPR2380]

Dodavatel: Abcam
AB79545-100 AB79545-1ML
ABCAAB79545-100EA 18530 CZK
ABCAAB79545-100 ABCAAB79545-1ML
Anti-delta 1 Catenin/CAS (phospho S268) Rabbit Monoclonal Antibody [clone: EPR2380]
Protilátky
Rabbit monoclonal [EPR2380] to delta 1 Catenin/CAS (phospho S268).

Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: delta 1 Catenin/CAS
Clonality: Monoclonal
Clone: EPR2380
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR