Tisk…

Anti-Metabotropic Glutamate Receptor 5 Rabbit Monoclonal Antibody [clone: EPR2425Y]

Dodavatel: Abcam
AB76316-100 AB76316-1ML
ABCAAB76316-100EA 19600 CZK
ABCAAB76316-100 ABCAAB76316-1ML
Anti-Metabotropic Glutamate Receptor 5 Rabbit Monoclonal Antibody [clone: EPR2425Y]
Protilátky
Rabbit monoclonal [EPR2425Y] to Metabotropic Glutamate Receptor 5.

Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Metabotropic Glutamate Receptor 5
Clonality: Monoclonal
Clone: EPR2425Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR